missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15678080 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15678080 500 g Poly bottle 500g
15668080 2.5 kg Poly bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15678080 Supplier Honeywell Research Chemicals Supplier No. S2252500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Melting Point 280°C
Color Colorless to Yellow
pH 8 to 9 (at 100.00g/L 20°C)
Quantity 500 g
Linear Formula NaNO2
UN Number UN1500
Formula Weight 69g/mol
Percent Purity ≥99.0%
Ignition Point 914°F
Odor Odorless
Packaging Poly bottle
Appearance Crystalline
Density 2.17g/cu.cm at 20°C
Chemical Name or Material Sodium nitrite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.