missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrite, 98%

Product Code. 11432958
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432958 1000 g 1000g
11442958 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432958 Supplier Thermo Scientific Alfa Aesar Supplier No. A18668.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7632-00-0 | NaNO2 | 69.00 g/mol

Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

Solubility
Soluble in water, methanol, ethanol, ammonia, nitric acid. Slightly soluble diethyl ether.

Notes
Decomposes above 320°. Hygroscopic. Incompatible with reducing agents, strong oxidizing agents, organics, finely powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NaNO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Melting Point 271°C (decomposition)
Quantity 1000 g
Linear Formula NaNO2
UN Number UN1500
Merck Index 14,8648
Solubility Information Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether.
Formula Weight 69
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.17
Chemical Name or Material Sodium nitrite
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium nitrite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.