missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, TraceSELECT™, ≥99.999% (metals basis), Honeywell™ Fluka™

Product Code. p-7120814 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15646840

Brand: Honeywell-Fluka 7175225G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point Decomposes on heating
Melting Point 308°C
Color Colorless
pH 5.0 to 7.0 (at 20°C)
Physical Form Solid
Quantity 25 g
Linear Formula NaNO3
UN Number UN1498
Solubility Information 883g/L (at 20°C)
Formula Weight 84.99g/mol
Percent Purity ≥99.999% (metals basis)
Grade TraceSELECT™
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Sodium nitrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.