missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, puriss. p.a. ACS Reagent, Solstice

Product Code. 15603430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
Unit Size:
100g
1kg
500g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15603430 500 g 500g
15683420 100 g 100g
15693420 1 kg 1kg
15623430 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15603430 Supplier Honeywell-Fluka Supplier No. 31440500G
  • ONLINE EXCLUSIVE PRICE
    £48.30
    £47.64 / 500g
    Save £0.66
    1% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Puriss. p.a., ACS Reagent, Reag. ISO,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium;nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Quantity 500 g
Linear Formula NaNO3
UN Number 1498
Formula Weight 84.99g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Nitrate, Reag. ISO
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.