missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, meets analytical spec. of FCC, E251, 99.0-100.5%, Solstice
BIOPHARMA

Product Code. 15661320
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
5kg
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661320 1 kg 1kg
15691320 5 kg 5kg
15601330 6 x 1 kg Pack of 6
15681320 4 x 5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15661320 Supplier Honeywell-Fluka Supplier No. 134441KG
  • ONLINE EXCLUSIVE PRICE
    £59.00
    £54.03 / 1kg
    Save £4.97
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets analytical specification of FCC, E251, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Physical Form Crystalline
Quantity 1 kg
Linear Formula NaNO3
UN Number 1498
Solubility Information Soluble in water 1gm/1.1mL, Soluble in alcohol 1gm/125mL, Soluble in boling water 1gm/0.6mL, Soluble in boiling alcohol 1gm/52mL
IUPAC Name sodium;nitrate
Formula Weight 84.99g/mol
Percent Purity 99 to 100.5%
Grade FCC
Chemical Name or Material Sodium Nitrate, Meets analytical specification of FCC
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.