Learn More
Sodium nitrate, extra pure, Thermo Scientific™
Brand: Thermo Scientific Acros 436090250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 7631-99-4 | |
| NNaO3 | |
| 15,8778 | |
| Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol | |
| [N+](=O)([O-])[O-].[Na+] | |
| 84.99 | |
| CHEBI:63005 | |
| ≥99.0% | |
| 2% max. (105°C) | |
| Sodium nitrate |
| 25kg | |
| NaNO3 | |
| sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
| VWDWKYIASSYTQR-UHFFFAOYSA-N | |
| sodium;nitrate | |
| 24268 | |
| 84.99 | |
| Extra Pure | |
| Fibre drum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.