missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, extra pure, Thermo Scientific™

Brand:  Thermo Scientific Acros 436090050

Product Code. 11924281

  • ONLINE EXCLUSIVE PRICE
    £169.00
    £164.00 / 5kg
    Save £5.00
    2% Off
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7631-99-4
NNaO3
15,8778
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
[N+](=O)([O-])[O-].[Na+]
84.99
CHEBI:63005
≥99.0%
2% max. (105°C)
Sodium nitrate
5kg
NaNO3
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
Extra Pure
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.