missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrate, Extra Pure, SLR, Fisher Chemical™
94.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
3 kg
Description
Sodium Nitrate, Extra Pure, SLR
Specifications
Boiling Point | 380°C |
Melting Point | 306°C |
Color | White |
pH | 5.5 to 8 |
Physical Form | Solid Crystalline |
Quantity | 1 kg |
IUPAC Name | sodium nitrate |
Formula Weight | 84.99g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium nitrate
- Warning
- Oxidising solid Category 3
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'