missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, ACS, 99.0% min

Product Code. 11352448
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
25g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352448 1 kg 1kg
11362448 25 g 25g
11342448 250 g 250g
11332448 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11352448 Supplier Thermo Scientific Alfa Aesar Supplier No. 014493.A1
  • ONLINE EXCLUSIVE PRICE
    £65.50
    £56.65 / 1kg
    Save £8.85
    13% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7631-99-4 ∣ NNaO3 ∣ 84.99 g/mol

In manufacturing of HNO{3}, as a catalyst to manufacture H{2}SO{4}, in manufacturing of glass, enamel for pottery, sodium nitriteSodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Melting Point 308°C
Physical Form Crystalline
Quantity 1 kg
Sensitivity Hygroscopic
Assay Percent Range 99.0% min.
Density 2.26
Solubility Information Soluble in water. Slightly soluble in methanol.
IUPAC Name sodium nitrate
Formula Weight 84.99
Percent Purity ≥99%
Grade ACS/Reagent
Chemical Name or Material Sodium nitrate, For ACS analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Sodium nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.