missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, ACS, 99.0% min, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 014493.30


Quantity 250g

 View more versions of this product

Product Code. 11342448

  • £30.10 / 250g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7631-99-4
84.994
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate
NNaO3
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]

Specifications

7631-99-4
99.0% min.
MFCD00011119
Soluble in water. Slightly soluble in methanol.
[N+](=O)([O-])[O-].[Na+]
84.994
CHEBI:63005
ACS Reagent
2.26
250g
NNaO3
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
Hygroscopic
Sodium nitrate, For ACS analysis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium nitrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contain: Sodium nitrate

SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.