Learn More
Sodium nitrate, 99.0% min., ACS, Alfa Aesar™
Brand: Alfa Aesar 014493.14
Code : NEW
Additional Details : CAS Number : 7631-99-4 Weight : 0.02500kg Transport : UN number : 1498 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
Sodium nitrate | |
7631-99-4 | |
99.0% min. | |
84.99 |
For ACS analysis | |
Crystalline | |
NaNO3 | |
MFCD00011119 |
Specifications
Sodium nitrate | |
7631-99-4 | |
99.0% min. | |
84.99 | |
2.26 |
For ACS analysis | |
Crystalline | |
NaNO3 | |
MFCD00011119 | |
25g |