missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 99.0% min., ACS, Alfa Aesar™

Brand:  Alfa Aesar 014493.A7

Code : NEW

Additional Details : CAS Number : 7631-99-4 Weight : 5.00000kg Transport : UN number : 1498 Chem class : 1 Pack group : III

Quantity 5kg

 View more versions of this product

Product Code. 11332448

  • £149.40 / 5kg


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium nitrate
7631-99-4
99.0% min.
84.99
For ACS analysis
Crystalline
NaNO3
MFCD00011119

Specifications

Sodium nitrate
7631-99-4
99.0% min.
84.99
2.26
For ACS analysis
Crystalline
NaNO3
MFCD00011119
5kg
Videos
SDS