missing translation for 'onlineSavingsMsg'
Learn More

Sodium nicotinate, 98%

Product Code. 11425866
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425866 500 g 500g
11415866 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425866 Supplier Thermo Scientific Alfa Aesar Supplier No. B22224.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54-86-4 | C6H4NNaO2 | 145.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 54-86-4
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00064352
InChI Key KFLRWGSAMLBHBV-UHFFFAOYSA-M
Synonym sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium
PubChem CID 23687810
IUPAC Name sodium;pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)[O-].[Na+]

Spezifikation

Melting Point >300°C
Quantity 500 g
Formula Weight 145.1
Percent Purity 98%
Chemical Name or Material Sodium nicotinate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.