missing translation for 'onlineSavingsMsg'
Learn More

Sodium Naphthalene-2,6-disulfonate, TRC

Product Code. 30420696
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420696 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420696 Supplier Toronto Research Chemicals Supplier No. TRCS64601010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Naphthalene-2,6-disulfonate
CAS 1655-45-4
Molecular Formula C10H6Na2O6S2
Quantity 10 g
Synonym 2,6-Naphthalenedisulfonic acid','sodium salt (1:2) (ACI), 2,6-Naphthalenedisulfonic acid','disodium salt (8CI','9CI), Disodium 2,6-naphthalenedisulfonate, Sodium naphthalene-2,6-disulfonate, 2,6-Naphthalenedisulfonic acid disodium salt
InChI Formula InChI=1S/C10H8O6S2.2Na/c11-17(12,13)9-3-1-7-5-10(18(14,15)16)4-2-8(7)6-9;;/h1-6H,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1ccc2cc(ccc2c1)S(=O)(=O)[O-]
IUPAC Name disodium;naphthalene-2,6-disulfonate
Molecular Weight (g/mol) 332.26
Formula Weight 331.9401
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.