missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-tridecyl sulfate, 99%, may cont. up to ca 5% water and residual solvent

Product Code. 11499587
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499587 25 g 25g
11479587 1 g 1g
11489587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499587 Supplier Thermo Scientific Alfa Aesar Supplier No. B22089.14
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3026-63-9 | C13H27NaO4S | 302.405 g/mol

Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.

Solubility
Soluble in water.

Notes
Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3026-63-9
Molecular Formula C13H27NaO4S
Molecular Weight (g/mol) 302.405
MDL Number MFCD00024993
InChI Key HQCFDOOSGDZRII-UHFFFAOYSA-M
Synonym sodium tridecyl sulfate, unii-jt30s80yr4, sodium n-tridecyl sulphate, sulfuric acid, mono-c8-18-alkyl esters, sodium salts, 1-tridecanol, 1-hydrogen sulfate , sodium salt 1:1, tridecyl sodium sulfate, sodium tridecyl sulphate, 1-tridecanol, hydrogen sulfate, sodium salt, sodium mono-c8-18-alkyl sulfates, sodium tridecylsulfate
PubChem CID 23668353
IUPAC Name sodium;tridecyl sulfate
SMILES CCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 185°C to 189°C
Linear Formula CH3(CH2)12OSO3Na
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 302.4
Percent Purity 99%
Assay may cont. up to ≈2% water and residual solvent
Chemical Name or Material Sodium n-tridecyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.