missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-octyl sulfate, 99%

Product Code. 11469363
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469363 25 g 25g
11449363 1 g 1g
11459363 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469363 Supplier Thermo Scientific Alfa Aesar Supplier No. A11786.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 142-31-4 | C8H17NaO4S | 232.27 g/mol

Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H17NaO4S
Molecular Weight (g/mol) 232.27
MDL Number MFCD00007470
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼195°C (decomposition)
Quantity 25 g
Beilstein 3658635
Solubility Information Soluble in water.
Formula Weight 232.28
Percent Purity 99%
Chemical Name or Material Sodium n-octyl sulfate
compliance-icons
Product Identifier
  • Sodium octyl sulphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium octyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.