missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-nonyl sulfate, 99%

Product Code. 11352516
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352516

Brand: Thermo Scientific Alfa Aesar L06418.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1072-15-7 | C9H19NaO4S | 246.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-15-7
Molecular Formula C9H19NaO4S
Molecular Weight (g/mol) 246.297
MDL Number MFCD00024893
InChI Key FTWCSAMTIKSPAT-UHFFFAOYSA-M
Synonym sodium nonyl sulfate, sodium nonyl sulphate, sodium n-nonyl sulfate, sulfuric acid, monononyl ester, sodium salt, unii-ft7kj97ok6, ft7kj97ok6, sulfuric acid, monononyl ester, sodium salt 1:1, sodium n-nonyl sulphate, dsstox_cid_27487, dsstox_rid_82377
PubChem CID 23677967
IUPAC Name sodium;nonyl sulfate
SMILES CCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Beilstein 3748751
Formula Weight 246.3
Percent Purity 99%
Chemical Name or Material Sodium n-nonyl sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.