missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-decyl sulfate, 98%

Product Code. 11447767
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447767 25 g 25g
11427767 1 g 1g
11437767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447767 Supplier Thermo Scientific Alfa Aesar Supplier No. A16299.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-87-0 | C10H21NaO4S | 260.324 g/mol

sodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.

Solubility
It is soluble in H20, poorly soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keepp away from acids Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-87-0
Molecular Formula C10H21NaO4S
Molecular Weight (g/mol) 260.324
MDL Number MFCD00041881
InChI Key XZTJQQLJJCXOLP-UHFFFAOYSA-M
Synonym sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
PubChem CID 23665771
IUPAC Name sodium;decyl sulfate
SMILES CCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Beilstein 3576860
Solubility Information It is soluble in H20,poorly soluble in organic solvents.
Formula Weight 260.33
Percent Purity 99%
Chemical Name or Material Sodium n-decyl sulfate
compliance-icons
Product Identifier
  • Sodium n-decyl sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium n-decyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.