missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium molybdenum oxide is widely used corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
Solubility
Soluble in water.
Notes
Stable. Incompatible with strong oxidizing agents.
Sodium molybdenum oxide is widely used corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
Solubility
Soluble in water.
Notes
Stable. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 100°C (in -2H2O) |
Quantity | 2500 g |
Merck Index | 14,8645 |
Solubility Information | Soluble in water. |
Formula Weight | 241.95 (205.92 Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Density | 2.71 |
Chemical Name or Material | Sodium molybdenum oxide dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'