missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdate(VI) dihydrate, ACS reagent

Product Code. 12357173 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 12357173

Brand: Thermo Scientific Acros 446365000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Na2MoO4·2H2O
Merck Index 15,8776
Solubility Information Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
Formula Weight 241.95
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 3.28
Density 3.2800g/mL
Chemical Name or Material Sodium molybdate(VI) dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.