missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdate, MedChemExpress

Product Code. 16481188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481188 Supplier Medchem Express Supplier No. HYD0851
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium molybdate (Molybdate disodium) is a useful source of molybdate.

Sodium molybdate (Molybdate disodium) is a useful source of molybdate.

Specifications

Chemical Name or Material Sodium molybdate
CAS 7631-95-0
Color White
Purity Grade Notes Research
Molecular Formula MoNa2O4
Quantity 5 g
Synonym Molybdate disodium
Solubility Information H2O : ≥ 100 mg/mL (485.58 mM)
SMILES O=[Mo](O[Na])(O[Na])=O
Molecular Weight (g/mol) 205.94
Formula Weight 205.94
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.