missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, puriss., Solstice
BIOPHARMA

Product Code. 15651290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15651290 2.5 kg 2.5kg
15641290 1 kg 1kg
15661290 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15651290 Supplier Honeywell-Fluka Supplier No. 133152.5KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph Eur, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Quantity 2.5 kg
Linear Formula Na2MoO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 241.95g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Molybdate Dihydrate, Meets analytical specification of PhEur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.