missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdate dihydrate, Honeywell Fluka™

Product Code. 15687920 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15687920 100 g 100g
15607930 500 g 500g
15697920 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15687920 Supplier Honeywell-Fluka Supplier No. M1003100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Quantity 100 g
Linear Formula Na2MoO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 241.95g/mol
Percent Purity ≥99.5%
Chemical Name or Material Sodium molybdate dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.