missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, ≥98%, Honeywell Fluka™

Product Code. 15622630 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Poly Bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15622630 5 g Poly Bottle 5g
15612630 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15622630 Supplier Honeywell-Fluka Supplier No. 2436555G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-95-0
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.926
MDL Number MFCD00003486
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
PubChem CID 61424
ChEBI CHEBI:75215
IUPAC Name disodium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 687°C
Color White
Quantity 5 g
Linear Formula Na2MoO4
UN Number NONH for all modes of transport
Formula Weight 205.92g/mol
Percent Purity ≥98%
Packaging Poly Bottle
Appearance Granular powder
Density 3.78g/mL at 25°C
Chemical Name or Material Sodium Molybdate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.