missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methylsulfinate, TRC

Product Code. 30423314
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423314 Supplier Toronto Research Chemicals Supplier No. TRCS6670705G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Methylsulfinate
CAS 20277-69-4
Molecular Formula CH3NaO2S
Quantity 5 g
Synonym Methanesulfinic Acid','Sodium Salt, Sodium Methanesulfinate
InChI Formula InChI=1S/CH4O2S.Na/c1-4(2)3;/h1H3,(H,2,3);/q;+1/p-1
SMILES [Na+].CS(=O)[O-]
IUPAC Name sodium;methanesulfinate
Molecular Weight (g/mol) 102.09
Formula Weight 101.9751
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.