missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Alkalinity | total alkalinity (as NaOCH3):> 99% |
Melting Point | 126.0°C |
Color | White-Yellow |
Density | 0.8 |
Packaging | Plastic Drum |
Linear Formula | CH3ONa |
Quantity | 2.5 kg |
UN Number | 1431 |
Beilstein | 01, 280 |
Fieser | 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352 |
Show More |
Safety and Handling
Product Identifier
- Sodium methoxide, anhydrous powder
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Substances and mixtures corrosive to metals Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H251-Self-heating: may catch fire.
- H290-May be corrosive to metals.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'