missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4089436
44.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
124-41-4, 67-56-1
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
124-41-4, 67-56-1
CH3NaO
54.02
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 65°C
Melting Point -98°C
Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula CH3ONa
Flash Point 11°C
Merck Index 15, 8775
Solubility Information Solubility in water: reacts
Formula Weight 54.02
Concentration or Composition (by Analyte or Components) 0.45 to 0.60M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.8
Density 0.8000g/mL
Chemical Name or Material Sodium methoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'