missing translation for 'onlineSavingsMsg'
Learn More

Sodium metavanadate, typically 96%, V 38% min

Product Code. 11346108
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346108 250 g 250g
11336108 25 g 25g
11356108 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346108 Supplier Thermo Scientific Alfa Aesar Supplier No. 011092.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13718-26-8 | NaO3V | 121.93 g/mol

Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-26-8
Molecular Formula NaO3V
Molecular Weight (g/mol) 121.93
MDL Number MFCD00011125
InChI Key CMZUMMUJMWNLFH-UHFFFAOYSA-N
Synonym sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
PubChem CID 4148882
ChEBI CHEBI:75221
SMILES [Na+].[O-][V](=O)=O

Specifications

Melting Point 630°C
Color White
Physical Form Chunks and/or Powder
Quantity 250 g
Assay Percent Range Typically 96%
Solubility Information Soluble in water.
IUPAC Name sodium vanadiumoylolate
Formula Weight 121.93
Odor Odorless
Appearance Off-white to light orange powder
Density 2.79g/cm3 at 20°C
Chemical Name or Material Sodium metavanadate, V 38% min.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.