Learn More
Sodium metatungstate monohydrate
Description
Sodium metatungstate monohydrate plays a vital role in sink and swim analysis. It is a derivative of sodium tungstate, which is a potential therapy for obesity. It acts as an inhibitor, which inhibits nucleoside-triphospahte- diphosphohydrolases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 2986.13 (2968.12 Anhydrous) |
| Odor | Odorless |
| Appearance | White powder |
| Density | 5.47 |
| Chemical Name or Material | Sodium metatungstate monohydrate |
Safety and Handling
- Sodium metatungstate monohydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Sodium metatungstate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.