missing translation for 'onlineSavingsMsg'
Learn More

Sodium metatungstate, MedChemExpress

Product Code. 16482866
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482866 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482866 Supplier Medchem Express Supplier No. HY103259
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium metatungstate (Sodium polyoxotungstate) is a potent ecto-nucleoside triphosphate diphosphohydrolase (ENTPDase) inhibitor, with Ki values of 2.58 μM, 3.26 μM, and 28.8 μM for NTPDase 1 (CD39), NTPDase 3 and NTPDase 2 respectively. Sodium metatungstate inhibits ATP breakdown but also blocks central synaptic transmission, an action independent of NTPDase inhibition.

Sodium metatungstate (Sodium polyoxotungstate) is a potent ecto-nucleoside triphosphate diphosphohydrolase (ENTPDase) inhibitor, with Ki values of 2.58 μM, 3.26 μM, and 28.8 μM for NTPDase 1 (CD39), NTPDase 3 and NTPDase 2 respectively. Sodium metatungstate inhibits ATP breakdown but also blocks central synaptic transmission, an action independent of NTPDase inhibition.

Specifications

Chemical Name or Material Sodium metatungstate
Physical Form Solid
CAS 12141-67-2
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula H2Na6O40W12
For Use With (Application) Neuroscience-Neuromodulation
Synonym Sodium polyoxotungstate POM-1
Solubility Information H2O : 50 mg/mL (16.74 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (16.74 mM; Need ultrasonic)
SMILES [3Na2WO4.9WO3]
Molecular Weight (g/mol) 2986.01
Formula Weight 2986.01
Percent Purity 93.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.