missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metatungstate Hydrate, TRC

Product Code. 30413505
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413505 5 g 5g
30412049 25 g 25g
30409997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413505 Supplier Toronto Research Chemicals Supplier No. TRCS6675005G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12141-67-2
Molecular Formula O39Na6W12•H2O
Molecular Weight (g/mol) 2986.01
Synonym Sodium metatungstate (Na6H2W12O40), Sodium tungstate (Na6H2W12O40), Sodium tungstate(VI) (Na6H2W12O40), Tungstate (W12(OH)2O386-) Hexasodium, Tungstic Acid (H8W12O40) Hexasodium Salt, SPT, POM 1
IUPAC Name hexasodium;dihydroxy(dioxo)tungsten;dioxido(dioxo)tungsten;trioxotungsten
SMILES OC(=O)c1ccc(cc1)NC[C@@H]2N(C=O)C3=C(NC(N)=NC3=O)NC2

Specifications

Quantity 5 g
InChI Formula InChI=1S/6Na.2H2O.38O.12W/h;;;;;;2*1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q6*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*-1;;;;;;;;;;;;+2/p-2
Formula Weight 2987.162
Chemical Name or Material Sodium Metatungstate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.