missing translation for 'onlineSavingsMsg'
Learn More

Sodium metatungstate hydrate, crystalline

Product Code. 10173281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173281 250 g Glass bottle 250g
10091601 50 g Glass bottle 50g
10423075 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10173281 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 304092500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 652154-12-6 | H2Na6O40W12·xH2O | 2986.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652154-12-6
Molecular Formula H2Na6O40W12·xH2O
Molecular Weight (g/mol) 2986.13
MDL Number MFCD00150663
Synonym Sodium polytungstate Hydrate

Specifications

Color White to Yellow
Physical Form Crystalline Powder or Beads
Quantity 250 g
Linear Formula Na6[H2W12O40]·xH2O
Solubility Information Solubility in water: >1000g/L (20°C)
Formula Weight 2986.13
Percent Purity ≥85% (WO3)
Packaging Glass bottle
Chemical Name or Material Sodium metatungstate hydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium metatungstate hydrate, crystalline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.