missing translation for 'onlineSavingsMsg'
Learn More

Sodium metatungstate, 2.82 density solution in water, stabilized

Product Code. 10603702 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
This item is not returnable. View return policy

Product Code. 10603702

Brand: Thermo Scientific Acros 304100500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12141-67-2 | H2Na6O40W12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12141-67-2
Molecular Formula H2Na6O40W12
MDL Number MFCD00150663
Synonym Sodium polytungstate

Specifications

Color Green to Yellow
Linear Formula Na6[H2W12O40]
Density 2.8200g/mL
Solubility Information Solubility in water: >1000g/L water (20°C).
Formula Weight 2986.13
Percent Purity ≥67% (WO3)
Packaging Glass bottle
Specific Gravity 2.82
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Sodium metatungstate, 2.82 Density Solution in Water, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium metatungstate, 2.82 density solution in water, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.