missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaborate tetrahydrate, 98%

Product Code. 11429393
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429393 10,000 g 10000g
11409393 500 g 500g
11419393 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429393 Supplier Thermo Scientific Alfa Aesar Supplier No. A11803.0C
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10555-76-7
Molecular Formula BH8NaO6
Molecular Weight (g/mol) 137.86
MDL Number MFCD00149205
InChI Key JAKYJVJWXKRTSJ-UHFFFAOYSA-N
Synonym sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
PubChem CID 23694267
IUPAC Name sodium;oxido(oxo)borane;tetrahydrate
SMILES O.O.O.O.[Na+].[O-]B=O

Specifications

Melting Point ∽53°C (decomposition)
Quantity 10,000 g
Formula Weight 137.86 (65.80 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Sodium metaborate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.