missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, MP Biomedicals™

Product Code. 11484642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
5 kg
Unit Size:
100g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484642 100 g 100g
11404652 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484642 Supplier MP Biomedicals Supplier No. 02191438.1
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Relied on by scientists. Sodium Metabisulfite, MP Biomedicals is ACS reagent grade with a purity greater than 97%. MP Biomedicals has delivered quality biochemicals for over 50 years.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 190.11g/mol
Percent Purity ≥97.0%
Grade ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.