missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, ≥97.0%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11494642 Shop All MP Biomedicals Products
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11494642 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494642 Supplier MP Biomedicals Supplier No. 0219143801
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7681-57-4
Quantity 1 kg
Assay Percent Range ≥97.0%
Molecular Formula Na2O5S2
MDL Number MFCD00167602
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O
IUPAC Name disodium (sulfinatooxy)sulfinate
Molecular Weight (g/mol) 190.09
PubChem CID 656671
Formula Weight 190.1
Grade ACS Reagent
Chemical Name or Material Sodium Metabisulfite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.