missing translation for 'onlineSavingsMsg'
Learn More

Sodium meta-Periodate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15172017
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Quantity 125 g
IUPAC Name sodium periodate
Formula Weight 213.89
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 3.87kg
Chemical Name or Material Sodium meta-Periodate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'