missing translation for 'onlineSavingsMsg'
Learn More

Sodium (meta)Arsenite, TRC

Product Code. 30418784
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418784 5 g 5g
30422524 10 g -
30413764 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418784 Supplier Toronto Research Chemicals Supplier No. TRCS0808685G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-46-5
Molecular Formula As O2 . Na
Molecular Weight (g/mol) 129.91
Synonym Sodium arsenate(III)
IUPAC Name sodium;dioxidoarsenic(3+)
SMILES [Na+].[O-][As]=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/AsHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1
Formula Weight 129.9001
Chemical Name or Material Sodium arsenite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.