missing translation for 'onlineSavingsMsg'
Learn More

Sodium (meta)arsenite concentrate, For 1L standard solution, 0.05 M NaAsO2, Honeywell Fluka™

Product Code. 15695230 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15695230

Brand: Honeywell-Fluka 381501EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 7784-46-5
Quantity 1 Pc.
Molecular Formula AsNaO2
Linear Formula NaAsO2
MDL Number MFCD00003472
UN Number UN1686
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES [O-][As]=O.[Na+]
IUPAC Name sodium;oxoarsinite
Molecular Weight (g/mol) 129.909
PubChem CID 443495
ChEBI CHEBI:29678
Formula Weight 129.91g/mol
Chemical Name or Material Sodium (meta)arsenite concentrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.