missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-(+)-tartrate dihydrate, ACS, 99.0-101.0%

Product Code. 11367548
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11367548

Brand: Thermo Scientific Alfa Aesar 033242.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

Sodium L-(+)-tartrate is used as an emulsifier, as a food additives, binding agent in food products like jellies, margarine and sausage casings. It is utilized as a common primary standard for Karl Fischer titration to measure the water content. It is also used as buffers for molecular biology and cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point ∽150°C-2H2O
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.0 - 101.0%
Beilstein 6121732
Solubility Information Soluble in waterSoluble in water. Insoluble in alcohol and organic solvents.
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Formula Weight 230.08 (194.06 Anhydrous)
Grade ACS
Density 1.82
Chemical Name or Material Sodium L-(+)-tartrate dihydrate, Suitable for Karl Fischer reagent standardization
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.