missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-(+)-tartrate dihydrate, 99%

Product Code. 11403267
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403267 1000 g 1000g
11413267 250 g 250g
11423267 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403267 Supplier Thermo Scientific Alfa Aesar Supplier No. A16187.0B
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point ∼150°C (in -2H2O)
Density 1.82
Odor Odorless
Quantity 1000 g
Beilstein 6121732
Merck Index 14,8684
Solubility Information It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents.
Formula Weight 230.08 (194.06 Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium L-(+)-tartrate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.