missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-(+)-Tartaric Acid Dihydrate, TRC

Product Code. 30012647
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012647 100 g 100g
30007247 10 g 10g
30003860 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012647 Supplier Toronto Research Chemicals Supplier No. TRCS673050100G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4 H4 O6 . 2 Na . 2 H2 O
Molecular Weight (g/mol) 230.08
Synonym Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, sodium salt, hydrate (1:2:2), Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, disodium salt, dihydrate (9CI), Butanedioic acid, 2,3-dihydroxy- [R-(R*,R*)]-, disodium salt, dihydrate, Tartaric acid, disodium salt, dihydrate (8CI), Disodium tartarate dihydrate, Disodium tartrate dihydrate, Sodium tartrate dihydrate
IUPAC Name disodium;(2R,3R)-2,3-dihydroxybutanedioate;dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C(=O)[O-])C(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1
Formula Weight 230.0015
Chemical Name or Material Sodium Tartrate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.