missing translation for 'onlineSavingsMsg'
Learn More

Sodium isopropoxide

Product Code. 11493770
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493770 100 g 100g
11483770 5 g 5g
11403780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493770 Supplier Thermo Scientific Alfa Aesar Supplier No. 089446.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 683-60-3 | C3H7NaO | 82.078 g/mol

Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-60-3
Molecular Formula C3H7NaO
Molecular Weight (g/mol) 82.078
MDL Number MFCD00061566
InChI Key WBQTXTBONIWRGK-UHFFFAOYSA-N
Synonym sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
PubChem CID 10920443
IUPAC Name sodium;propan-2-olate
SMILES CC(C)[O-].[Na+]

Specifications

Melting Point 70°C to 75°C
Color White
Linear Formula NaOCH(CH3)2
Quantity 100 g
UN Number UN3206
Sensitivity Air and moisture sensitive
Solubility Information Soluble in tetrahydrofuran.
Formula Weight 82.08
Physical Form Crystalline
Chemical Name or Material Sodium isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.