missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, TRC

Product Code. 30012032
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012032 10 g 10g
30004832 5 g 5g
30007124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012032 Supplier Toronto Research Chemicals Supplier No. TRCS63800010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
Synonym Anayodin, Ioduril, NSC 77388, Sodium Monoiodide, Soiodin;
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/HI.Na/h1H;/q;+1/p-1
Formula Weight 149.8942
Chemical Name or Material Sodium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.