missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide Symporter Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Sodium Iodide Symporter
Concentration 0.77 mg/mL
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16862805
View Documents
Proteintech
24324-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16852805
View Documents
Proteintech
24324-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.

Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extra membranous domains. It may represent an autoantigen in thyroid.
Specifications

Specifications

Sodium Iodide Symporter
Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human, Rat
Q63008, Q92911, Q99PN0
114479, 114613, 6528
Sodium iodide symporter Fusion Protein Ag19504
Primary
-20°C
SLC5A5
0.77 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
NIS, SLC5A5, Sodium iodide symporter, Sodium/iodide cotransporter
SLC5A5
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.