missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11367027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367027 5 g 5g
11387027 25 g 25g
11377027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367027 Supplier Thermo Scientific Alfa Aesar Supplier No. 010871.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003532

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Solubility Information Soluble in water (1840mg/ml at 25°C),alcohol,glycerol,and acetone. Insoluble in ether.
Formula Weight 149.89 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Sodium iodide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.