missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide dihydrate, 99+%, for analysis

Product Code. 10414935 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10414935 500 g Glass bottle 500g
10032740 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414935 Supplier Thermo Scientific Acros Supplier No. 202895000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13517-06-1 | INa·2H2O | 185.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-06-1
Molecular Formula INa·2H2O
Molecular Weight (g/mol) 185.93
MDL Number MFCD00149211
InChI Key VEEIYHUOLCUPSA-UHFFFAOYSA-M
Synonym sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
PubChem CID 23685997
IUPAC Name sodium;iodide;dihydrate
SMILES O.O.[Na+].[I-]

Specifications

Melting Point 752°C
Color White
pH 6 to 9 (5% aq. soln.)
Physical Form Adhering Fine Crystals
Quantity 500 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NaI·2H2O
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 14,89
Merck Index 10, 8469
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,glycerol and acetone,very soluble in nh3
Formula Weight 185.93
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Specific Gravity 2.44
Density 2.4400g/mL
Chemical Name or Material Sodium iodide dihydrate, For analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium iodide dihydrate
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium iodide dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.