missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, ACS, ≥99.5%, Honeywell™ Fluka™

Product Code. 15646970 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 15646970

Brand: Honeywell-Fluka 746371100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Free-flowing, ACS reagent, anhydrous,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Melting Point 662°C
Color Colorless
Physical Form Solid
Quantity 100 g
Linear Formula NaI
UN Number UN3077
Solubility Information 1.793g/L at 20°c
Formula Weight 149.89g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Density 3.67g/cm3; Vapor Density: >1 (vs air)
Chemical Name or Material Sodium iodide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.