Learn More
Sodium iodide, 99+% (dry wt., water <1.5%)
Description
Sodium iodide is widely used for halide exchange (Finkelstein reaction), for example in the conversion of an alkyl chloride, allyl chloride and arylmethyl chloride into their respective iodides, which are precursors for pharmaceutical and fine chemical products. They are used to ehnance the efficiency of the formation of Wittig adducts from less reactive chlorides and bromides. Appropriate prepartions find use as a nutrient supplement. Sodium iodide is used as the precursor to the control agent in ab initio emulsion polymerization. Sodium iodide finds use in the determination of dissolved oxygen in the modified Winkler method, synthesis of the fluorescent dye coppersensor-1 (CS1) for imaging labile copper pools in biological samples, and the cleavage of esters, lactones, carbamates and ethers in combination with chlorotrimethylsilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium iodide is widely used for halide exchange (Finkelstein reaction), for example in the conversion of an alkyl chloride, allyl chloride and arylmethyl chloride into their respective iodides, which are precursors for pharmaceutical and fine chemical products. They are used to ehnance the efficiency of the formation of Wittig adducts from less reactive chlorides and bromides. Appropriate prepartions find use as a nutrient supplement. Sodium iodide is used as the precursor to the control agent in ab initio emulsion polymerization. Sodium iodide finds use in the determination of dissolved oxygen in the modified Winkler method, synthesis of the fluorescent dye coppersensor-1 (CS1) for imaging labile copper pools in biological samples, and the cleavage of esters, lactones, carbamates and ethers in combination with chlorotrimethylsilane.
Solubility
Very soluble in water and liquid ammonia. Freely soluble in acetone, formic acid, formamide, methanol, and acetonitrile. Soluble in N,N-dimethylformamide.
Notes
Deliquescent. Store in a tightly closed container. Incompatible with strong oxidizing agents and acids.
Specifications
Specifications
| Boiling Point | 1300°C to 1304°C |
| Melting Point | 651°C |
| Quantity | 100 g |
| Merck Index | 14,8631 |
| Solubility Information | Very soluble in water and liquid ammonia. Freely soluble in acetone,formic acid,formamide,methanol,and acetonitrile. Soluble in N,N-dimethylformamide. |
| Formula Weight | 149.89 |
| Percent Purity | ≥99% |
| Odor | Odorless |
| Refractive Index | 1.7745 |
| Sensitivity | Hygroscopic; Light sensitive |
| Show More |
Safety and Handling
- Sodium iodide
- Danger
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contain: Sodium iodide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.