missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium iodate, 99%, extra pure, reagent, powder, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 560.0°C |
Color | White |
pH | 5.0 to 8.0 (5% soln. at 20°C) |
Physical Form | Fine Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 98.5% min. (Iodometry) |
Linear Formula | NaIO3 |
Merck Index | 15, 8764 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol |
Formula Weight | 197.89 |
Show More |
Safety and Handling
Product Identifier
- Sodium iodate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'